α'-Bromo-α,α,α-trifluoro-o-xylene 98%
Supplier: Apollo Scientific
Danger
PC7631-100G PC7631-25G PC7631-5G
APOSPC7631-100GEA 98.6 EUR
APOSPC7631-100G APOSPC7631-25G APOSPC7631-5G
α'-Bromo-α,α,α-trifluoro-o-xylene 98%
α'-Bromo-α,α,α-trifluoro-o-xylene
Formula: C₈H₆BrF₃
Boiling Pt: 70…72 °C/7mmHg
Melting Pt: 27…30 °C
Density: 1.571 g/cm³ (25 °C)
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00013558
CAS Number: 395-44-8
EINECS: 206-901-7
UN: 3261
ADR: 8,II