α'-Chloro-α,α,α-trifluoro-p-xylene 98%
Supplier: Apollo Scientific
Danger
Synonyms: 4-(Chloromethyl)benzotrifluoride, 1-(Chloromethyl)-4-(trifluoromethyl)benzene, 1-(Chlormethyl)-4-(trifluormethyl)benzene, 4-(Trifluoromethyl)benzyl chloride
PC7633G-25G PC7633G-5G
APOSPC7633G-25GEA 37.7 EUR
APOSPC7633G-25G APOSPC7633G-5G
α'-Chloro-α,α,α-trifluoro-p-xylene 98%
α'-Chloro-α,α,α-trifluoro-p-xylene
Formula: C₈H₆ClF₃
MW: 194.58 g/mol
Boiling Pt: 68 °C/12mmHg
Melting Pt: 19 °C
Density: 1.315 g/cm³ (25 °C)
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00040772
CAS Number: 939-99-1
UN: 3265
ADR: 8 II