2-Bromo-6-(trifluoromethyl)pyridine 98%
Supplier: Apollo Scientific
Danger
PC7657-100G PC7657-25G PC7657-5G
APOSPC7657-100GEA 369 EUR
APOSPC7657-100G APOSPC7657-25G APOSPC7657-5G
2-Bromo-6-(trifluoromethyl)pyridine 98%
2-Bromo-6-(trifluoromethyl)pyridine
Formula: C₆H₃BrF₃N
MW: 226 g/mol
Boiling Pt: 78…79 °C/14mmHg
Melting Pt: 48…52 °C
Storage Temperature: Ambient
MDL Number: MFCD00153087
CAS Number: 189278-27-1
UN: 2811
ADR: 6.1 III