4-(Trifluoromethyl)cinnamic acid 98%
Supplier: Apollo Scientific
Warning
Synonyms: trans-4-(Trifluoromethyl)cinnamic acid
PC7664-100G PC7664-25G PC7664-500G
APOSPC7664-100GEA 74.1 EUR
APOSPC7664-100G APOSPC7664-25G APOSPC7664-500G
4-(Trifluoromethyl)cinnamic acid 98%
4-(Trifluoromethyl)cinnamic acid
Formula: C₁₀H₇F₃O₂
MW: 216.16 g/mol
Melting Pt: 230…234 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002696
CAS Number: 16642-92-5