3-(Trifluoromethyl)crotonic acid 95%
Supplier: Apollo Scientific
Danger
PC7668-25G PC7668-5G PC7668-100MG
APOSPC7668-25GEA 487 EUR
APOSPC7668-25G APOSPC7668-5G APOSPC7668-100MG
3-(Trifluoromethyl)crotonic acid 95%
3-(Trifluoromethyl)crotonic acid
Formula: C₅H₅F₃O₂
MW: 154.09 g/mol
Boiling Pt: 158…163 °C
Melting Pt: 28…30 °C
Density: 1.318 g/cm³ (25 °C)
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00014374
CAS Number: 69056-67-3
UN: 3261
ADR: 8 III