4-(Trifluoromethyl)piperidine hydrochloride
Supplier: Apollo Scientific
Warning
PC7713-1G PC7713-5G PC7713-10G
APOSPC7713-1GEA 21.9 EUR
APOSPC7713-1G APOSPC7713-5G APOSPC7713-10G
4-(Trifluoromethyl)piperidine hydrochloride
4-(Trifluoromethyl)piperidine hydrochloride
Formula: C₆H₁₁ClF₃N
MW: 189.61 g/mol
Melting Pt: 155…160 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD04971993
CAS Number: 155849-49-3