3-Bromo-4-(trifluoromethyl)pyridine
Supplier: Apollo Scientific
Warning
PC7718-25G PC7718-5G
APOSPC7718-25GEA 400 EUR
APOSPC7718-25G APOSPC7718-5G
3-Bromo-4-(trifluoromethyl)pyridine
3-Bromo-4-(trifluoromethyl)pyridine
Formula: C₆H₃BrF₃N
MW: 226 g/mol
Boiling Pt: 58 °C/11mmHg
Storage Temperature: Ambient
MDL Number: MFCD08061310
CAS Number: 936841-70-2
UN: 2810
ADR: 6.1 III