4-(Trifluoromethyl)-2,3,5,6-tetrafluorothiophenol 97%
Supplier: Apollo Scientific
Warning
PC7769C-1G PC7769C-5G
APOSPC7769C-1GEA 71.2 EUR
APOSPC7769C-1G APOSPC7769C-5G
4-(Trifluoromethyl)-2,3,5,6-tetrafluorothiophenol 97%
4-(Trifluoromethyl)-2,3,5,6-tetrafluorothiophenol
Formula: C₇HF₇S
Boiling Pt: 169 °C
Density: 1.66 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00191594
CAS Number: 651-84-3