4-(Trifluoromethyl)thiobenzamide 98%
Supplier: Apollo Scientific
Warning
PC7769F-25G PC7769F-5G PC7769F-100G
APOSPC7769F-25GEA 34.8 EUR
APOSPC7769F-25G APOSPC7769F-5G APOSPC7769F-100G
4-(Trifluoromethyl)thiobenzamide 98%
4-(Trifluoromethyl)thiobenzamide
Formula: C₈H₆F₃NS
Melting Pt: 135…138 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00051806
CAS Number: 72505-21-6
UN: 2811
ADR: 6.1,III