1,2,4-Trifluoro-5-nitrobenzene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2,4,5-Trifluoronitrobenzene
PC7790-100G PC7790-250G PC7790-1KG
APOSPC7790-100GEA 65.4 EUR
APOSPC7790-100G APOSPC7790-250G APOSPC7790-1KG
1,2,4-Trifluoro-5-nitrobenzene 98%
1,2,4-Trifluoro-5-nitrobenzene
Formula: C₆H₂F₃NO₂
Boiling Pt: 194…195 °C/760mmHg
Melting Pt: –11 °C
Density: 1.544 g/cm³ (25 °C)
Flash Pt: 89 °C
Storage Temperature: Ambient
MDL Number: MFCD00007051
CAS Number: 2105-61-5
EINECS: 218-281-5
UN: 2810
ADR: 6.1,III