5-(3,5-Difluorophenyl)isoxazole
Supplier: Apollo Scientific
Warning
PC7829-1G PC7829-250MG
APOSPC7829-1GEA 58 EUR
APOSPC7829-1G APOSPC7829-250MG
5-(3,5-Difluorophenyl)isoxazole
5-(3,5-Difluorophenyl)isoxazole
Formula: C₉H₅F₂NO
Melting Pt: 75…79 °C
Storage Temperature: Ambient
MDL Number: MFCD05664750
CAS Number: 874800-58-5