3-Bromo-2,6-difluorobenzaldehyde 98%
Supplier: Apollo Scientific
Warning
PC7849-25G PC7849-5G PC7849-100G
APOSPC7849-25GEA 130 EUR
APOSPC7849-25G APOSPC7849-5G APOSPC7849-100G
3-Bromo-2,6-difluorobenzaldehyde 98%
3-Bromo-2,6-difluorobenzaldehyde
Formula: C₇H₃BrF₂O
Melting Pt: 53…57 °C
Storage Temperature: Ambient
MDL Number: MFCD08282773
CAS Number: 398456-82-1