2-Amino-4,4,4-trifluoro-3-methylbutanoic acid 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4,4,4-Trifluoro-DL-valine
PC7855-250MG
PC7855-100MG
APOSPC7855-250MGEA
613
EUR
APOSPC7855-250MG
APOSPC7855-100MG
2-Amino-4,4,4-trifluoro-3-methylbutanoic acid 97%
2-Amino-4,4,4-trifluoro-3-methylbutanoic acid
Formula:
C₅H₈F₃NO₂ MW: 171.12 g/mol Melting Pt: 245…248 °C Storage Temperature: Ambient |
MDL Number:
MFCD00021721 CAS Number: 16063-79-9 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...