tert-Butyl-4-chloro-2-fluorobenzoate 97%
Supplier: Apollo Scientific
PC7916-1G PC7916-5G PC7916-25G
APOSPC7916-1GEA 128 EUR
APOSPC7916-1G APOSPC7916-5G APOSPC7916-25G
tert-Butyl-4-chloro-2-fluorobenzoate 97%
tert-Butyl 4-chloro-2-fluorobenzoate
Formula: C₁₁H₁₂ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD09475908
CAS Number: 941294-14-0