Tris[4-(trifluoromethyl)phenyl]phosphine 95+%
Supplier: Apollo Scientific
Synonyms: Tris(4-trifluoromethylphenyl)phosphine, Tris(4-(trifluoromethyl)phenyl)phosphine
PC7919G-1G PC7919G-5G PC7919G-10G
APOSPC7919G-1GEA 36.4 EUR
APOSPC7919G-1G APOSPC7919G-5G APOSPC7919G-10G
Tris[4-(trifluoromethyl)phenyl]phosphine 95+%
Tris[4-(trifluoromethyl)phenyl]phosphine
Formula: C₂₁H₁₂F₉P
MW: 466.28 g/mol
Melting Pt: 73…76 °C
Storage Temperature: Ambient
MDL Number: MFCD00058883
CAS Number: 13406-29-6