2,6-Dichloro-4-(trifluoromethyl)pyridine 98%
Supplier: Apollo Scientific
Danger
PC7947-1G PC7947-25G PC7947-5G
APOSPC7947-1GEA 21.9 EUR
APOSPC7947-1G APOSPC7947-25G APOSPC7947-5G
2,6-Dichloro-4-(trifluoromethyl)pyridine 98%
2,6-Dichloro-4-(trifluoromethyl)pyridine
Formula: C₆H₂Cl₂F₃N
Boiling Pt: 56…57 °C/10mmHg
Density: 1.505 g/cm³ (25 °C)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00042246
CAS Number: 39890-98-7
UN: 2810
ADR: 6.1,III