ω-Fluoroacetophenone
Supplier: Apollo Scientific
Danger
Synonyms: 2-Fluoro-1-phenylethanone, Phenacyl fluoride
PC7958-1G PC7958-250MG
APOSPC7958-1GEA 87 EUR
APOSPC7958-1G APOSPC7958-250MG
ω-Fluoroacetophenone
ω-Fluoroacetophenone
Formula: C₈H₇FO
Boiling Pt: 61.5…64.4 °C/1.3mmHg
Melting Pt: 26…27 °C
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD08461612
CAS Number: 450-95-3
UN: 1759
ADR: 8,III