2-[(Trifluoromethyl)sulphanyl]aniline 98%
Supplier: Apollo Scientific
Warning
PC8023-25G PC8023-5G
APOSPC8023-25GEA 372 EUR
APOSPC8023-25G APOSPC8023-5G
2-[(Trifluoromethyl)sulphanyl]aniline 98%
2-[(Trifluoromethyl)sulphanyl]aniline
Formula: C₇H₆F₃NS
MW: 193.19 g/mol
Melting Pt: 30…34 °C
Flash Pt: 90.55 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00236333
CAS Number: 347-55-7
EINECS: 206-473-1