4-(Trifluoromethyl)-1H-indazole
Supplier: Apollo Scientific
Warning
PC8056-1G PC8056-250MG PC8056-5G
APOSPC8056-1GEA 150 EUR
APOSPC8056-1G APOSPC8056-250MG APOSPC8056-5G
4-(Trifluoromethyl)-1H-indazole
4-(Trifluoromethyl)-1H-indazole
Formula: C₈H₅F₃N₂
Melting Pt: 107…109 °C
Storage Temperature: Ambient
MDL Number: MFCD09263211
CAS Number: 1000339-98-9