4-Chloro-3-fluorobenzotrifluoride 99%
Supplier: Apollo Scientific
Warning
PC8215-5G PC8215-1G PC8215-25G
APOSPC8215-5GEA 43.5 EUR
APOSPC8215-5G APOSPC8215-1G APOSPC8215-25G
4-Chloro-3-fluorobenzotrifluoride 99%
4-Chloro-3-fluorobenzotrifluoride
Formula: C₇H₃ClF₄
MW: 198.55 g/mol
Boiling Pt: 135 °C
Density: 1.49 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00060646
CAS Number: 32137-20-5
UN: 1993
ADR: 3 III