4-Bromo-2,6-difluorobenzaldehyde
Supplier: Apollo Scientific
Warning
PC8236-25G PC8236-5G
APOSPC8236-25GEA 37.7 EUR
APOSPC8236-25G APOSPC8236-5G
4-Bromo-2,6-difluorobenzaldehyde
4-Bromo-2,6-difluorobenzaldehyde
Formula: C₇H₃BrF₂O
Melting Pt: 76…81 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD03094459
CAS Number: 537013-51-7
UN: 2811
ADR: 6.1,III