2,6-Difluoro-4-methoxybenzoyl chloride 97%
Supplier: Apollo Scientific
Danger
PC8313-10G PC8313-1G PC8313-5G
APOSPC8313-10GEA 200 EUR
APOSPC8313-10G APOSPC8313-1G APOSPC8313-5G
2,6-Difluoro-4-methoxybenzoyl chloride 97%
2,6-Difluoro-4-methoxybenzoyl chloride
Formula: C₈H₅ClF₂O₂
MW: 206.58 g/mol
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD03094495
CAS Number: 125369-56-4
UN: 3265
ADR: 8 III