1-Bromo-2-fluorocyclohexane 97%
Supplier: Apollo Scientific
Warning
PC8345-1G PC8345-250MG PC8345-5G
APOSPC8345-1GEA 241 EUR
APOSPC8345-1G APOSPC8345-250MG APOSPC8345-5G
1-Bromo-2-fluorocyclohexane 97%
1-Bromo-2-fluorocyclohexane
Formula: C₆H₁₀BrF
Boiling Pt: 74…78 °C/19mmHg
Flash Pt: 56.8 °C
Storage Temperature: Ambient
MDL Number: MFCD01320789
CAS Number: 656-57-5
UN: 1993
ADR: 3,III