1,2-Dichloro-4-fluoro-5-nitrobenzene 98%
Supplier: Apollo Scientific
Warning
PC8376-25G PC8376-5G PC8376-100G
APOSPC8376-25GEA 46.4 EUR
APOSPC8376-25G APOSPC8376-5G APOSPC8376-100G
1,2-Dichloro-4-fluoro-5-nitrobenzene 98%
1,2-Dichloro-4-fluoro-5-nitrobenzene
Formula: C₆H₂Cl₂FNO₂
Boiling Pt: 247 °C
Melting Pt: 17 °C
Density: 1.596 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00075330
CAS Number: 2339-78-8