2-Bromo-2'-fluoro-4'-(trifluoromethyl)acetophenone
Supplier: Apollo Scientific
Danger
Synonyms: 2-Fluoro-4-(trifluoromethyl)phenacyl bromide
PC8406-1G PC8406-5G PC8406-25G
APOSPC8406-1GEA 52.2 EUR
APOSPC8406-1G APOSPC8406-5G APOSPC8406-25G
2-Bromo-2'-fluoro-4'-(trifluoromethyl)acetophenone
2-Bromo-2'-fluoro-4'-(trifluoromethyl)acetophenone
Formula: C₉H₅BrF₄O
MW: 285.0330 g/mol
Melting Pt: 30…37 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03094505
CAS Number: 537050-12-7
UN: 1759
ADR: 8 III