2-Bromo-4'-fluoro-2'-(trifluoromethyl)acetophenone
Supplier: Apollo Scientific
Danger
Synonyms: 4-Fluoro-2-(trifluoromethyl)phenacyl bromide
PC8409-1G PC8409-250MG PC8409-5G
APOSPC8409-1GEA 140 EUR
APOSPC8409-1G APOSPC8409-250MG APOSPC8409-5G
2-Bromo-4'-fluoro-2'-(trifluoromethyl)acetophenone
2-Bromo-4'-fluoro-2'-(trifluoromethyl)acetophenone
Formula: C₉H₅BrF₄O
Boiling Pt: 62…64 °C/0.06mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD03094508
CAS Number: 537050-13-8
UN: 1760
ADR: 8,III