2-[Cyano(phenyl)methyl]-6-fluorobenzenecarbonitrile
Supplier: Apollo Scientific
Warning
Synonyms: 2-(2-Cyano-3-fluorophenyl)-2-phenylacetonitrile
PC8445-1G PC8445-250MG
APOSPC8445-1GEA 143 EUR
APOSPC8445-1G APOSPC8445-250MG
2-[Cyano(phenyl)methyl]-6-fluorobenzenecarbonitrile
2-[Cyano(phenyl)methyl]-6-fluorobenzenecarbonitrile
Formula: C₁₅H₉FN₂
MW: 236.25 g/mol
Melting Pt: 80…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00231973
CAS Number: 338791-66-5