3-[5-(Trifluoromethyl)-2-pyridinyl]aniline
Supplier: Apollo Scientific
Warning
PC8644-1G PC8644-250MG
APOSPC8644-1GEA 434 EUR
APOSPC8644-1G APOSPC8644-250MG
3-[5-(Trifluoromethyl)-2-pyridinyl]aniline
3-[5-(Trifluoromethyl)-2-pyridinyl]aniline
Formula: C₁₂H₉F₃N₂
Melting Pt: 84…86 °C
Storage Temperature: Ambient
MDL Number: MFCD08276968
CAS Number: 886361-00-8