4-Chloro-1,1,1-trifluorobutane 97%

Supplier: Apollo Scientific
Warning

PC8645-1G PC8645-5G
APOSPC8645-1GEA 110 EUR
APOSPC8645-1G APOSPC8645-5G
4-Chloro-1,1,1-trifluorobutane 97%
4-Chloro-1,1,1-trifluorobutane
Formula: C₄H₆ClF₃
Boiling Pt: 81 °C
Melting Pt: –67.25 °C
Density: 1.2425 g/cm³ (25 °C)
Storage Temperature: Ambient
CAS Number: 406-85-9
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR