1-Bromo-3-fluoro-4-(trifluoromethoxy)benzene 97%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Bromo-2-fluoro-1-(trifluoromethoxy)benzene
PC8727-5G PC8727-25G PC8727-100G
APOSPC8727-5GEA 21.9 EUR
APOSPC8727-5G APOSPC8727-25G APOSPC8727-100G
1-Bromo-3-fluoro-4-(trifluoromethoxy)benzene 97%
1-Bromo-3-fluoro-4-(trifluoromethoxy)benzene
Formula: C₇H₃BrF₄O
MW: 259.00 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01861132
CAS Number: 105529-58-6