4-Nitrophenylsulphur pentafluoride

Supplier: Apollo Scientific
Danger

PC8766-1G PC8766-25G PC8766-5G
APOSPC8766-1GEA 348 EUR
APOSPC8766-1G APOSPC8766-25G APOSPC8766-5G
4-Nitrophenylsulphur pentafluoride
4-Nitrophenylsulphur pentafluoride
Formula: C₆H₄F₅NO₂S
Boiling Pt: 76…77 °C/12mmHg
Melting Pt: 36…38 °C
Storage Temperature: Ambient
MDL Number: MFCD00221619
CAS Number: 2613-27-6
EINECS: 447-610-7
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR