5-Fluoro-2-(trifluoromethyl)benzoyl chloride 97%
Supplier: Apollo Scientific
Danger
PC8790-25G PC8790-5G PC8790-100G
APOSPC8790-25GEA 214 EUR
APOSPC8790-25G APOSPC8790-5G APOSPC8790-100G
5-Fluoro-2-(trifluoromethyl)benzoyl chloride 97%
5-Fluoro-2-(trifluoromethyl)benzoyl chloride
Formula: C₈H₃ClF₄O
Boiling Pt: 168…169 °C
Density: 1.5 g/cm³ (25 °C)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00083534
CAS Number: 216144-70-6
UN: 3265
ADR: 8,II