2-Bromo-3,5-difluorophenol 96%
Supplier: Apollo Scientific
Warning
PC8835-10G PC8835-1G PC8835-5G
APOSPC8835-10GEA 299 EUR
APOSPC8835-10G APOSPC8835-1G APOSPC8835-5G
2-Bromo-3,5-difluorophenol 96%
2-Bromo-3,5-difluorophenol
Formula: C₆H₃BrF₂O
MW: 208.99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01861127
CAS Number: 325486-43-9