2-Fluoro-m-toluidine
Supplier: Apollo Scientific
Warning
PC8849-5G PC8849-25G PC8849-100G
APOSPC8849-5GEA 21.9 EUR
APOSPC8849-5G APOSPC8849-25G APOSPC8849-100G
2-Fluoro-m-toluidine
2-Fluoro-m-toluidine
Formula: C₇H₈FN
Density: 1.1 g/cm³ (25 °C)
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD06410915
CAS Number: 1978-33-2