2-Piperidin-1-yl-5-(trifluoromethyl)pyridine
Supplier: Apollo Scientific
PC8992-1G PC8992-100MG PC8992-250MG PC8992-500MG PC8992-2.5G PC8992-5G
APOSPC8992-1GEA 393 EUR
APOSPC8992-1G APOSPC8992-100MG APOSPC8992-250MG APOSPC8992-500MG APOSPC8992-2.5G APOSPC8992-5G
2-Piperidin-1-yl-5-(trifluoromethyl)pyridine
2-Piperidin-1-yl-5-(trifluoromethyl)pyridine
Formula: C₁₁H₁₃F₃N₂
Storage Temperature: Ambient
MDL Number: MFCD00547912
CAS Number: 132664-24-5