3-Ethoxy-2-fluorobenzoic acid 98%
Supplier: Apollo Scientific
PC901467-1G PC901467-5G
APOSPC901467-1GEA 256 EUR
APOSPC901467-1G APOSPC901467-5G
3-Ethoxy-2-fluorobenzoic acid 98%
3-Ethoxy-2-fluorobenzoic acid
Formula: C₉H₉FO₃
MW: 184.1600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10699646
CAS Number: 1033201-71-6