N-Ethyl-4-fluorobenzamide 98%
Supplier: Apollo Scientific
PC901487-100G
PC901487-25G
PC901487-5G
APOSPC901487-100GEA
899
EUR
APOSPC901487-100G
APOSPC901487-25G
APOSPC901487-5G
N-Ethyl-4-fluorobenzamide 98%
N-Ethyl 4-fluorobenzamide
Formula:
C₉H₁₀FNO Storage Temperature: Ambient |
MDL Number:
MFCD01214153 CAS Number: 772-18-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...