3-Fluoro-N-isopropylbenzamide 95%

Supplier: Apollo Scientific

Synonyms: N-Isopropyl 3-fluorobenzamide

PC901492-100G PC901492-25G PC901492-5G
APOSPC901492-100GEA 632 EUR
APOSPC901492-100G APOSPC901492-25G APOSPC901492-5G
3-Fluoro-N-isopropylbenzamide 95%
3-Fluoro-N-isopropylbenzamide
Formula: C₁₀H₁₂FNO
Storage Temperature: Ambient
MDL Number: MFCD01341858
CAS Number: 459218-95-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR