4-Chloro-5-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine ≥98%
Supplier: Apollo Scientific
PC909023-100MG PC909023-1G PC909023-500MG
APOSPC909023-100MGEA 487 EUR
APOSPC909023-100MG APOSPC909023-1G APOSPC909023-500MG
4-Chloro-5-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine ≥98%
4-Chloro-5-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine
Formula: C₈H₄ClF₃N₂
Storage Temperature: Ambient
MDL Number: MFCD15529110
CAS Number: 1196507-58-0