3-Fluoro-4-methoxypyridine 97%
Supplier: Apollo Scientific
PC910743-1G PC910743-5G
APOSPC910743-1GEA 128 EUR
APOSPC910743-1G APOSPC910743-5G
3-Fluoro-4-methoxypyridine 97%
3-Fluoro-4-methoxypyridine
Formula: C₆H₆FNO
Storage Temperature: Ambient
MDL Number: MFCD13185605
CAS Number: 1060805-03-9