3-Fluoro-4-(trifluoromethyl)benzaldehyde 97%
Supplier: Apollo Scientific
Warning
PC9127-1G PC9127-25G PC9127-5G
APOSPC9127-1GEA 21.9 EUR
APOSPC9127-1G APOSPC9127-25G APOSPC9127-5G
3-Fluoro-4-(trifluoromethyl)benzaldehyde 97%
3-Fluoro-4-(trifluoromethyl)benzaldehyde
Formula: C₈H₄F₄O
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00236275
CAS Number: 204339-72-0