4-Bromo-2,3-difluorobenzaldehyde 95+%
Supplier: Apollo Scientific
Warning
PC9172-25G PC9172-5G
APOSPC9172-25GEA 201 EUR
APOSPC9172-25G APOSPC9172-5G
4-Bromo-2,3-difluorobenzaldehyde 95+%
4-Bromo-2,3-difluorobenzaldehyde
Formula: C₇H₃BrF₂O
MW: 221.00 g/mol
Melting Pt: 61…65 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09258953
CAS Number: 644985-24-0