4'-Methyl-3'-(trifluoromethyl)acetophenone 97%
Supplier: Apollo Scientific
PC9188-1G PC9188-5G PC9188-10G
APOSPC9188-1GEA 116 EUR
APOSPC9188-1G APOSPC9188-5G APOSPC9188-10G
4'-Methyl-3'-(trifluoromethyl)acetophenone 97%
4'-Methyl-3'-(trifluoromethyl)acetophenone
Formula: C₁₀H₉F₃O
MW: 202.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08532499
CAS Number: 851262-98-1