4-Bromo-3-(trifluoromethyl)phenylacetic acid
Supplier: Apollo Scientific
Warning
PC9210-1G PC9210-5G
APOSPC9210-1GEA 195 EUR
APOSPC9210-1G APOSPC9210-5G
4-Bromo-3-(trifluoromethyl)phenylacetic acid
4-Bromo-3-(trifluoromethyl)phenylacetic acid
Formula: C₉H₆BrF₃O₂
Melting Pt: 92…95 °C
Storage Temperature: Ambient
CAS Number: 914637-17-5