4-Amino-3-methylbenzotrifluoride 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Methyl-4-(trifluoromethyl)aniline
PC9254-10G PC9254-1G PC9254-5G
APOSPC9254-10GEA 608 EUR
APOSPC9254-10G APOSPC9254-1G APOSPC9254-5G
4-Amino-3-methylbenzotrifluoride 97%
4-Amino-3-methylbenzotrifluoride
Formula: C₈H₈F₃N
Boiling Pt: 98…99 °C/13mmHg
Melting Pt: 20…25 °C
Density: 1.2 g/cm³ (25 °C)
Flash Pt: 84 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03701068
CAS Number: 67169-22-6
UN: 2810
ADR: 6.1,III