2-(Trifluoromethyl)nicotinonitrile
Supplier: Apollo Scientific
Warning
PC9261-1G PC9261-250MG
APOSPC9261-1GEA 203 EUR
APOSPC9261-1G APOSPC9261-250MG
2-(Trifluoromethyl)nicotinonitrile
2-(Trifluoromethyl)nicotinonitrile
Formula: C₇H₃F₃N₂
MW: 172.11 g/mol
Density: 1.3473 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD08741366
CAS Number: 870066-15-2