Methyl 5-Cyano-1,6-Dihydro-1-(4-Fluorophenyl)-4-Methyl-6-Pyridazinone-3-Carboxylate 97%
Supplier: Apollo Scientific
PC9404-1G PC9404-500MG
APOSPC9404-1GEA 799 EUR
APOSPC9404-1G APOSPC9404-500MG
Methyl 5-Cyano-1,6-Dihydro-1-(4-Fluorophenyl)-4-Methyl-6-Pyridazinone-3-Carboxylate 97%
Methyl-5-cyano-1,6-dihydro-1-(4-fluorophenyl)-4-methyl-6-pyridazinone-3-carboxylate
Formula: C₁₄H₁₀FN₃O₃
MW: 287.25 g/mol
Melting Pt: 163…165 °C
Storage Temperature: Ambient
MDL Number: MFCD01315976
CAS Number: 339018-13-2