Methyl-4-hydroxy-6-oxo-1-[3-(trifluoromethyl)phenyl]-1,6-dihydro-3-pyridazinecarboxylate 97%
Supplier: Apollo Scientific
Warning
PC9416-1G PC9416-250MG
APOSPC9416-1GEA 256 EUR
APOSPC9416-1G APOSPC9416-250MG
Methyl-4-hydroxy-6-oxo-1-[3-(trifluoromethyl)phenyl]-1,6-dihydro-3-pyridazinecarboxylate 97%
Methyl-4-hydroxy-6-oxo-1-[3-(trifluoromethyl)phenyl]-1,6-dihydro-3-pyridazinecarboxylate
Formula: C₁₃H₉F₃N₂O₄
MW: 314.22 g/mol
Melting Pt: 119…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00141006
CAS Number: 121582-55-6