Methyl 5-(2-methoxy-2-oxoethyl)-1-[3-(trifluoro-methyl)phenyl]-1H-1,2,3-triazole-4-carboxylate 97%
Supplier: Apollo Scientific
PC9417-1G PC9417-500MG PC9417-5G
APOSPC9417-1GEA 275 EUR
APOSPC9417-1G APOSPC9417-500MG APOSPC9417-5G
Methyl 5-(2-methoxy-2-oxoethyl)-1-[3-(trifluoro-methyl)phenyl]-1H-1,2,3-triazole-4-carboxylate 97%
Methyl-5-(2-methoxy-2-oxoethyl)-1-[3-(trifluoro-methyl)phenyl]-1H-1,2,3-triazole-4-carboxylate
Formula: C₁₄H₁₂F₃N₃O₄
MW: 343.26 g/mol
MDL Number: MFCD00138717
CAS Number: 303995-97-3