[1-Methyl-3-(trifluoromethyl)-1H-thieno-[2,3-c]pyrazol-5-yl]methanol
Supplier: Apollo Scientific
PC9460-1G PC9460-500MG
APOSPC9460-1GEA 924 EUR
APOSPC9460-1G APOSPC9460-500MG
[1-Methyl-3-(trifluoromethyl)-1H-thieno-[2,3-c]pyrazol-5-yl]methanol
[1-Methyl-3-(trifluoromethyl)-1H-thieno-[2,3-c]pyrazol-5-yl]methanol
Formula: C₈H₇F₃N₂OS
MW: 236.22 g/mol
Melting Pt: 88…92 °C
Storage Temperature: Ambient
MDL Number: MFCD01936006
CAS Number: 338982-22-2